Skip to Content
Merck
(Z)-9-Tricosene 97%

(Z)-9-Tricosene

(Z)-9-Tricosene 97%
Synonym(s):
cis-9-Tricosene, Muscalure
Linear Formula:
CH3(CH2)12CH=CH(CH2)7CH3
CAS Number:
Molecular Weight:
322.61
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing