![Withanoside IV phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/137/321/88b71cee-afc4-48bd-8987-c3ef8e462c6d/640/88b71cee-afc4-48bd-8987-c3ef8e462c6d.png)
Withanoside IV
![Withanoside IV phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/137/321/88b71cee-afc4-48bd-8987-c3ef8e462c6d/640/88b71cee-afc4-48bd-8987-c3ef8e462c6d.png)
Synonym(s):
(1α,3β,22R)-3-[(6-O-β-D-Glucopyranosyl-β-D-glucopyranosyl)oxy]-1,22,27-trihydroxy-ergosta-5,24-dien-26-oic acid δ-lactone
Empirical Formula (Hill Notation):
C40H62O15
CAS Number:
Molecular Weight:
782.91
Products
Formula Weight
Manufacturer
Physical Form
Product Line
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing