Skip to Content
Merck
Withaferin A analytical standard

Withaferin A

Withaferin A analytical standard
Synonym(s):
5,6-Epoxy-4,27-dihydroxy-1-oxowitha-2,24-dienolide, Withaferine A, 4β,5β,6β,22R)-5,6-Epoxy-4,22,27-trihydroxy-1-oxo-ergosta-2,24-dien-26-oic acid δ-lactone, Withaferine
Empirical Formula (Hill Notation):
C28H38O6
CAS Number:
Molecular Weight:
470.60

Products

Product Category
Brand
Biological Source
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Format
Quality Segment
Functional Group
Application
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing