Skip to Content
AR
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Whirl-Pak
®
Blender Bag
Products
Product Category
bags (2)
Manufacturer
Capacity (mL)
Length (cm)
Color
Sterility
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
WPB01474
Whirl-Pak
®
Blender Bag,
capacity 5441 mL (184 oz), bag W × L × thickness 37.5 cm × 50 cm × 0.102 mm
Expand
View Pricing
WPB01421
Whirl-Pak
®
Blender Bag,
capacity 1627 mL (55 oz), bag W × L × thickness 19 cm × 30 cm × 0.076 mm
Expand
View Pricing