<iframe src="https://www.googletagmanager.com/ns.html?id=GTM-TXMJ2GG>m_auth=>m_preview=>m_cookies_win=x" height="0" width="0" style="display:none;visibility:hidden" id="tag-manager"></iframe>
Skip to Content
Products
Cart
0
AR
EN
Products
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Login
Order Lookup
Quick Order
Cart
0
Back
Whatman
®
Mini-UniPrep
®
G2 standard septum
Products
Product Category
filters (10)
syringe filters (1)
Color
Manufacturer
Cap
Color
Housing Material
Membrane Material
Vial Bottle Color
Package Size
Pore size (µm)
Sterility
Filter Products
Showing 1-10 of 10
Compare
Product Number
Description
Pricing
GN203NPERC
Whatman
®
Mini-UniPrep
®
G2 standard septum,
regenerated cellulose membrane, pore size 0.2 μm, clear vial, pack of 100
Expand
View Pricing
GN203APEORG
Whatman
®
Mini-UniPrep
®
G2 standard septum,
PTFE membrane, pore size 0.2 μm, amber vial, pack of 100
Expand
View Pricing
WHAGN203NPUORG
Whatman
®
Mini-UniPrep
®
G2 standard septum,
PTFE membrane, pore size 0.45 μm, clear vial color, pack of 100
Expand
View Pricing
WHAGN203NPEORG
Whatman
®
Mini-UniPrep
®
G2 standard septum,
PTFE membrane, pore size 0.2 μm, clear vial color, pack of 100
Expand
View Pricing
GN203APEAQU
Whatman
®
Mini-UniPrep
®
G2 standard septum,
PVDF membrane, pore size 0.2 μm, amber vial, pack of 100
Expand
View Pricing
WHAGN203NPUAQU
Whatman
®
Mini-UniPrep
®
G2 standard septum,
PVDF membrane, pore size 0.45 μm, clear vial color, pack of 100
Expand
View Pricing
GN503NPEORG
Whatman
®
Mini-UniPrep
®
G2 standard septum,
PTFE membrane, pore size 0.2 μm, clear vial, pack of 1000
Expand
View Pricing
WHAGN203NPEAQU
Whatman
®
Mini-UniPrep
®
G2 standard septum,
PVDF membrane, pore size 0.2 μm, clear vial color, pack of 100
Expand
View Pricing
GN203NPENYL
Whatman
®
Mini-UniPrep
®
G2 standard septum,
Nylon membrane, pore size 0.2 μm, clear vial, pack of 100
Expand
View Pricing
GN203NPUGMF
Whatman
®
Mini-UniPrep
®
G2 standard septum,
glass microfiber membrane, pore size 0.45 μm, clear vial, pack of 100
Expand
View Pricing
Choose up to 4 products to compare
Compare
Clear All
An unknown error has occured.
Español - ES
English - EN
Learn More