Skip to Content
AR
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Vinyl-Coated Lead Ring ("C" shape)
Products
Manufacturer
Color
Package Size
Filter Products
Showing 1-8 of 8
Compare
Product Number
Description
Pricing
HS8882A
Vinyl-Coated Lead Ring ("C" shape),
fits beaker size, 125 to 500 mL, blue
Expand
View Pricing
HS8882B
Vinyl-Coated Lead Ring ("C" shape),
fits beaker size, 250 to 1000 mL, blue
Expand
View Pricing
HS8881D
Vinyl-Coated Lead Ring ("C" shape),
fits beaker size, 1000 to 4000 mL, blue
Expand
View Pricing
HS8882C
Vinyl-Coated Lead Ring ("C" shape),
fits beaker size, 500 to 2000 mL, blue
Expand
View Pricing
HS8881A
Vinyl-Coated Lead Ring ("C" shape),
fits beaker size, 125 to 500 mL, blue
Expand
View Pricing
HS8881B
Vinyl-Coated Lead Ring ("C" shape),
fits beaker size, 250 to 1000 mL, blue
Expand
View Pricing
HS8882D
Vinyl-Coated Lead Ring ("C" shape),
fits beaker size, 1000 to 4000 mL, blue
Expand
View Pricing
HS8881C
Vinyl-Coated Lead Ring ("C" shape),
fits beaker size, 500 to 2000 mL, blue
Expand
View Pricing