Vicine
Synonym(s):
2,6-Diamino-5-(β-D-glucopyranosyloxy)-4(1H)-pyrimidinone, Divicine 5-(β-D-glucopyranoside, NSC 95092, Vicioside
Empirical Formula (Hill Notation):
C10H16N4O7
CAS Number:
Molecular Weight:
304.26
Products
Formula Weight
Manufacturer
Physical Form
Product Line
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing