Skip to Content
Merck
Vasoactive Intestinal Peptide human, porcine, rat ≥95% (HPLC), powder

Vasoactive Intestinal Peptide human, porcine, rat

Vasoactive Intestinal Peptide human, porcine, rat ≥95% (HPLC), powder
Synonym(s):
VIP
Empirical Formula (Hill Notation):
C147H238N44O42S
CAS Number:
Molecular Weight:
3325.80

Products

Product Category
Brand
Biological Source
Formula Weight
Gene Alias
Physical Form
Purity
Quality Segment
Showing 1-2 of 2
Compare
Product Number
Description
Pricing