![Vasicine European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/185/876/f7ec0f6c-d371-4b8b-8ef1-4a6738d24cb2/640/f7ec0f6c-d371-4b8b-8ef1-4a6738d24cb2.png)
Vasicine
![Vasicine European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/185/876/f7ec0f6c-d371-4b8b-8ef1-4a6738d24cb2/640/f7ec0f6c-d371-4b8b-8ef1-4a6738d24cb2.png)
Synonym(s):
Vasicine, (−)-Peganine, (3S)-1,2,3,9-Tetrahydropyrrolo[2,1-b]quinazolin-3-ol
Empirical Formula (Hill Notation):
C11H12N2O
CAS Number:
Molecular Weight:
188.23
Products
Formula Weight
Manufacturer
Application
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing