Skip to Content
Merck
Valspodar ≥98% (HPLC)

Valspodar

Valspodar ≥98% (HPLC)
Synonym(s):
6-[(2S,4R,6E)-4-Methyl-2-(methylamino)-3-oxo-6-octenoic acid]cyclosporin D, Amdray, PSC833, [3′-Desoxy-3′-oxo-MeBmt]1-[Val]2-cyclosporin
Empirical Formula (Hill Notation):
C63H111N11O12
CAS Number:
Molecular Weight:
1214.62

Products

Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Research Area
Showing 1-1 of 1
Compare
Product Number
Description
Pricing