Skip to Content
Merck
Valproic Acid Impurity K Pharmaceutical Secondary Standard; Certified Reference Material

Valproic Acid Impurity K

Valproic Acid Impurity K Pharmaceutical Secondary Standard; Certified Reference Material
Synonym(s):
(2RS)-2-Ethyl-2-methylpentanoic acid, 2-Ethyl-2-methylvaleric acid, NSC 466
Empirical Formula (Hill Notation):
C8H16O2
CAS Number:
Molecular Weight:
144.21

Products

Brand
Formula Weight
Physical Form
Agency
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1