Skip to Content
Merck
Ultrafiltration Discs, 30 kDa NMW Ultracel®, filter diam. 76 mm

Ultrafiltration Discs, 30 kDa NMW

Ultrafiltration Discs, 30 kDa NMW Ultracel®, filter diam. 76 mm
Synonym(s):
Regenerated Cellulose Membrane Filter, 30 kDa NMWCO

Products

Brand
Diameter (mm)
Surface Area cm²
Quality Segment
Membrane Material
MWCO (kDa)
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing