
TSKgel® BioAssist® Phenyl (10 µm) HPLC Columns

Synonym(s):
TSKgel® Hydrophobic Interaction Chromatography (HIC) Column
Products
Product Category
Manufacturer
Product Line
Separation Technique
ID (mm)
Length (cm)
Particle Size (µm)
Pore Size (Å)
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing