![TSKgel® Alpha-2500 HPLC Column phase hydroxylated methacrylate, L × I.D. 30 cm × 7.8 mm, 7 μm particle size](/deepweb/assets/sigmaaldrich/product/images/178/311/7143e77f-5e0a-416f-a205-ff5c7aec04e3/640/7143e77f-5e0a-416f-a205-ff5c7aec04e3.jpg)
TSKgel® Alpha-2500 (7 µm) HPLC Columns
![TSKgel® Alpha-2500 HPLC Column phase hydroxylated methacrylate, L × I.D. 30 cm × 7.8 mm, 7 μm particle size](/deepweb/assets/sigmaaldrich/product/images/178/311/7143e77f-5e0a-416f-a205-ff5c7aec04e3/640/7143e77f-5e0a-416f-a205-ff5c7aec04e3.jpg)
Synonym(s):
TSKgel® Alpha Columns
Products
Product Category
Manufacturer
Product Line
Agency
USP Suitability
Separation Technique
ID (mm)
Length (cm)
Particle Size (µm)
Pore Size (Å)
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing