![Tritosulfurone Metabolite M635H004 certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland](/deepweb/assets/sigmaaldrich/product/structures/426/104/0f8f3af9-da80-4080-9473-e5b1da11a09e/640/0f8f3af9-da80-4080-9473-e5b1da11a09e.png)
Tritosulfurone Metabolite M635H004
![Tritosulfurone Metabolite M635H004 certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland](/deepweb/assets/sigmaaldrich/product/structures/426/104/0f8f3af9-da80-4080-9473-e5b1da11a09e/640/0f8f3af9-da80-4080-9473-e5b1da11a09e.png)
Synonym(s):
2-Amino-4-methoxy-6-(trifluoromethyl)-1,3,5-triazine
Empirical Formula (Hill Notation):
C5H5F3N4O
CAS Number:
Molecular Weight:
194.11
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing