Skip to Content
AR
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Tris(2-cyanoethyl)nitromethane
Synonym(s):
4-(2-Cyanoethyl)-4-nitropimelonitrile, 3,3′,3″-(Nitromethylidyne)tripropionitrile, 4-(2-Cyanoethyl)-4-nitroheptanedinitrile, Nitro-tris(2-cyanoethyl)methane, Nitromethanetrispropionitrile
Linear Formula:
NO
2
C(CH
2
CH
2
CN)
3
CAS Number:
1466-48-4
Molecular Weight:
220.23
EC Number:
215-983-3
Products
Product Category
building blocks (1)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Filter Products
Showing 1-1 of 1
Compare
Product Number
Description
Pricing
357944
Tris(2-cyanoethyl)nitromethane,
95%
Expand
View Pricing