Skip to Content
Merck
(Trimethylsilyl)methacrylate 98%

(Trimethylsilyl)methacrylate

(Trimethylsilyl)methacrylate 98%
Linear Formula:
H2C=C(CH3)CO2Si(CH3)3
CAS Number:
Molecular Weight:
158.27

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Monomer Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing