![Triflumizole PESTANAL®, analytical standard](/deepweb/assets/sigmaaldrich/product/structures/281/044/2ce10407-f5ae-4e2c-9713-966a1334d807/640/2ce10407-f5ae-4e2c-9713-966a1334d807.png)
Triflumizole
![Triflumizole PESTANAL®, analytical standard](/deepweb/assets/sigmaaldrich/product/structures/281/044/2ce10407-f5ae-4e2c-9713-966a1334d807/640/2ce10407-f5ae-4e2c-9713-966a1334d807.png)
Synonym(s):
(E)-4-Chloro-α,α,α-trifluoro-N-(1-imidazol-1-yl-2-propoxyethylidene)-o-toluidine, 1-{1-{[4-Chloro-2-(trifluoromethyl)phenyl]imino}-2-propoxyethyl}-1H-imidazole
Empirical Formula (Hill Notation):
C15H15ClF3N3O
CAS Number:
Molecular Weight:
345.75
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2