Triethylphosphine solution
Synonym(s):
PEt3, Triethylphosphorous solution
Linear Formula:
(C2H5)3P
CAS Number:
Molecular Weight:
118.16
Products
Brand
Formula Weight
Physical Form
Functional Group
Markush Class
Markush Group
Reaction Type
Reagent Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing