Triethylamine trihydrofluoride
Synonym(s):
Hydrogen fluoride triethylamine, TREAT-HF
Linear Formula:
(C2H5)3N · 3HF
Products
Brand
Boiling Point (°C)
Formula Weight
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing