Skip to Content
Merck
Triethoxy(octyl)silane 97%

Triethoxy(octyl)silane

Triethoxy(octyl)silane 97%
Synonym(s):
Octyltriethoxysilane, Triethoxyoctylsilane
Linear Formula:
CH3(CH2)7Si(OC2H5)3
CAS Number:
Molecular Weight:
276.49

Products

Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-3 of 3
Compare
Product Number
Description
Pricing