Triamcinolone hexacetonide
Synonym(s):
(11β,16α)-21-(3,3-Dimethyl-1-oxobutoxy)-9-fluoro-11-hydroxy-16,17-[(1-methylethylidene)bis(oxy)]pregna-1,4-diene-3,20-dione, Triamcinolone hexacetonide
Empirical Formula (Hill Notation):
C30H41FO7
CAS Number:
Molecular Weight:
532.64
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2