Skip to Content
Merck
Triamcinolone Acetonide Related Compound C certified reference material, pharmaceutical secondary standard

Triamcinolone acetonide Related Compound C

Triamcinolone Acetonide Related Compound C certified reference material, pharmaceutical secondary standard
Synonym(s):
(11β,16α)-9-Fluoro-11,21,21-trihydroxy-16,17-[(1-methylethylidene)bis(oxy)]pregna-1,4-diene-3,20-dione
Empirical Formula (Hill Notation):
C24H31FO7
CAS Number:
Molecular Weight:
450.50

Products

Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade