Skip to Content
AR
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
trans
-3-Methyl-2-pentene
Empirical Formula (Hill Notation):
C
6
H
12
CAS Number:
616-12-6
Molecular Weight:
84.16
EC Number:
210-465-3
Products
Product Category
building blocks (1)
Brand
Sigma-Aldrich (1)
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Special Grade
Purum (1)
Filter Products
Showing 1-1 of 1
Compare
Product Number
Description
Pricing
68490
trans
-3-Methyl-2-pentene,
≥98.0% (GC)
Expand
View Pricing