
TOYOPEARL® AF-rProtein A HC-650F Bulk Media

Synonym(s):
TOYOPEARL® AF-rProtein A HC-650F Bulk Media
Products
Manufacturer
pH Value
Physical Form
Product Line
Separation Technique
Package Size
Particle Size (µm)
Pore Size (Å)
Technique
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
TOYOPEARL® AF-rProtein A HC-650F Bulk Media, phase rProtein A, bottle of 100 mL, 45 μm particle size