
TOYOPEARL® AFFIPAK ACT Sampler (AF-Epoxy, Tresyl-650M)

Synonym(s):
TOYOPEARL® AFFIPAK Activated Media Sampler
Products
Manufacturer
Physical Form
Product Line
Separation Technique
Package Size
Particle Size (µm)
Pore Size (Å)
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing