Skip to Content
AR
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
TLC aluminum sheets, Silica gel 60 F₂₅₄
Products
Product Category
sheets (5)
Brand
Supelco (5)
Length (cm)
Quality Segment
Fluorescence
Matrix Material
Support Material
Particle Size (µm)
Pore Size (Å)
Technique
Filter Products
Showing 1-5 of 5
Compare
Product Number
Description
Pricing
1.05554
TLC aluminum sheets, Silica gel 60 F₂₅₄,
pkg of 25 sheets, sheet L × W 20 cm × 20 cm, aluminum support
Expand
View Pricing
1.05549
TLC aluminum sheets, Silica gel 60 F₂₅₄,
pkg of 20 sheets, sheet L × W 5 cm × 7.5 cm, aluminum support
Expand
View Pricing
1.16834
TLC aluminum sheets, Silica gel 60 F₂₅₄,
pkg of 50 sheets, sheet L × W 5 cm × 10 cm, aluminum support
Expand
View Pricing
1.16717
TLC aluminum sheets, Silica gel 60 F₂₅₄,
pkg of 100 sheets, sheet L × W 10 cm × 10 cm, aluminum support
Expand
View Pricing
1.05534
TLC aluminum sheets, Silica gel 60 F₂₅₄,
pkg of 100 sheets, sheet L × W 5 cm × 20 cm, aluminum support
Expand
View Pricing