Skip to Content
AR
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Titanium/Molybdenum
Products
Manufacturer
Physical Form
Length (m)
Length (mm)
Filter Products
Showing 1-7 of 7
Compare
Product Number
Description
Pricing
GF23276426
Titanium/Molybdenum,
wire, Ti 85%/Mo 15%, 1.0 mm diameter, length 1 m, temper as drawn
Expand
View Pricing
GF29748796
Titanium/Molybdenum,
rod, Ti 85%/Mo 15%, 2.0 mm diameter, length 1000 mm, temper annealed
Expand
View Pricing
GF47752386
Titanium/Molybdenum,
rod, Ti 85%/Mo 15%, 2.0 mm diameter, length 100 mm, temper annealed
Expand
View Pricing
GF09552708
Titanium/Molybdenum,
wire, Ti 85%/Mo 15%, 1.0 mm diameter, length 2 m, temper as drawn
Expand
View Pricing
GF68556167
Titanium/Molybdenum,
rod, 2.0 mm diameter, Ti85%/Mo15%, length 500 mm, temper annealed
Expand
View Pricing
GF89190467
Titanium/Molybdenum,
rod, 2.0 mm diameter, Ti85%/Mo15%, length 200 mm, temper annealed
Expand
View Pricing
GF78586217
Titanium/Molybdenum,
wire, Ti85%/Mo15%, 1.0 mm diameter, length 0.5 m, temper as drawn
Expand
View Pricing