Skip to Content
Merck
Timolol maleate salt ≥98% (TLC), powder

Timolol maleate salt

Timolol maleate salt ≥98% (TLC), powder
Synonym(s):
S-(−)-1-(t-Butylamino)-3-[(4-morpholino-1,2,5-thiadiazol-3-yl)oxy]-2-propanol maleate salt, Timolol hydrogen maleate
Empirical Formula (Hill Notation):
C13H24N4O3S · C4H4O4
CAS Number:
Molecular Weight:
432.49
EC Number:

Products

Product Category
Brand
Color
Formula Weight
Gene Alias
Melting Point (°C)
Physical Form
Purity
Research Area
Showing 1-1 of 1
Compare
Product Number
Description
Pricing