Skip to Content
AR
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Thiotepa
Synonym(s):
Thio-TEPA, N,N′,N″-Triethylenethiophosphoramide, Thio-Tep, Thiofozil, Thiotef, Tio-tef, Tiofosfamid, Tiofosyl, Tiofozil, Tris(aziridinyl)phosphine sulfide
Empirical Formula (Hill Notation):
C
6
H
12
N
3
PS
CAS Number:
52-24-4
Molecular Weight:
189.22
EC Number:
200-135-7
Products
Product Category
antibiotics (1)
Brand
Sigma-Aldrich (1)
Color
Formula Weight
Manufacturer
Physical Form
Activity Spectrum
Format
Application
Mode of Action
API Family
Reference Material Type
Special Grade
pharmaceutical primary standard (1)
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
1664000
Thiotepa,
United States Pharmacopeia (USP) Reference Standard
Expand
View Pricing
T6069
Thiotepa,
Expand
View Pricing