
Thiamazole impurity C

Synonym(s):
1-Methyl-2-(methylthio)-1H-imidazole, 1-Methyl-2-(methylsulphanyl)-1H-imidazole
Empirical Formula (Hill Notation):
C5H8N2S
CAS Number:
Molecular Weight:
128.20
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing