Skip to Content
Merck
Tetrazolium Blue Chloride used in colorimetric determination of reducing compounds

Tetrazolium Blue Chloride

Tetrazolium Blue Chloride used in colorimetric determination of reducing compounds
Synonym(s):
3,3′-(3,3′-Dimethoxy[1,1′-biphenyl]-4,4′-diyl)-bis(2,5-diphenyl-2H-tetrazolium) dichloride, 3,3′-Dianisole-4,4′-bis(3,5-diphenyltetrazolium chloride), BTC, Blue Tetrazolium chloride, TZ
Empirical Formula (Hill Notation):
C40H32Cl2N8O2
CAS Number:
Molecular Weight:
727.64
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Application
Showing 1-2 of 2
Compare
Product Number
Description
Pricing