![Tetrabutylammonium phosphate monobasic solution 1.0 M in H2O](/deepweb/assets/sigmaaldrich/product/structures/312/057/04820820-19ad-44a6-9dc2-de869632c8dd/640/04820820-19ad-44a6-9dc2-de869632c8dd.png)
Tetrabutylammonium phosphate monobasic solution
![Tetrabutylammonium phosphate monobasic solution 1.0 M in H2O](/deepweb/assets/sigmaaldrich/product/structures/312/057/04820820-19ad-44a6-9dc2-de869632c8dd/640/04820820-19ad-44a6-9dc2-de869632c8dd.png)
Synonym(s):
Tetrabutylammonium dihydrogen phosphate
Linear Formula:
(CH3CH2CH2CH2)4N[OP(OH)2O]
CAS Number:
Molecular Weight:
339.45
Products
Brand
Formula Weight
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing