tert-Butyldichlorophosphine solution
Synonym(s):
(1,1-Dimethylethyl)phosphonous dichloride solution
Empirical Formula (Hill Notation):
C4H9Cl2P
CAS Number:
Molecular Weight:
158.99
Products
Brand
Formula Weight
Physical Form
Quality Segment
Functional Group
Markush Class
Markush Group
Reaction Type
Reagent Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing