Skip to Content
Merck
Terbutaline hemisulfate salt

Terbutaline hemisulfate salt

Terbutaline hemisulfate salt
Synonym(s):
2-t-Butylamino-1-(3,5-dihydroxyphenyl)ethanol, 5-[2-[(1,1-Dimethylethyl)amino]-1-hydroxyethyl]-1,3-benzenediol hemisulfate salt
Linear Formula:
C12H19NO3 · 1/2H2SO4
CAS Number:
Molecular Weight:
274.32
EC Number:

Products

Product Category
Brand
Formula Weight
Gene Alias
Research Area
Showing 1-1 of 1
Compare
Product Number
Description
Pricing