Skip to Content
AR
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Synthware
™
pressure equalizing funnel with high vacuum valve
Products
Product Category
funnels (6)
Manufacturer
Capacity (mL)
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
SYNF644125
Synthware
™
pressure equalizing funnel with high vacuum valve,
125 mL, joint: ST/NS 24/40, valve size 0-4 mm
Expand
View Pricing
SYNF642460
Synthware
™
pressure equalizing funnel with high vacuum valve,
60 mL, joint: ST/NS 24/40, valve size 0-4 mm
Expand
View Pricing
SYNF641425
Synthware
™
pressure equalizing funnel with high vacuum valve,
25 mL, joint: ST/NS 14/20, valve size 0-4 mm
Expand
View Pricing
SYNF641960
Synthware
™
pressure equalizing funnel with high vacuum valve,
60 mL, joint: ST/NS 19/22, valve size 0-4 mm
Expand
View Pricing
SYNF641460
Synthware
™
pressure equalizing funnel with high vacuum valve,
60 mL, joint: ST/NS 14/20, valve size 0-4 mm
Expand
View Pricing
SYNF641125
Synthware
™
pressure equalizing funnel with high vacuum valve,
125 mL, joint: ST/NS 14/20, valve size 0-4 mm
Expand
View Pricing