Skip to Content
Merck
Suramin sodium salt ≥98% (TLC)

Suramin sodium salt

Suramin sodium salt ≥98% (TLC)
Synonym(s):
Suramine sodium salt
Empirical Formula (Hill Notation):
C51H34N6Na6O23S6
CAS Number:
Molecular Weight:
1429.17
EC Number:

Products

Product Category
Color
Formula Weight
Gene Alias
Physical Form
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing