Strychnine
Synonym(s):
(−)-Strychnine
Empirical Formula (Hill Notation):
C21H22N2O2
CAS Number:
Molecular Weight:
334.41
Products
Formula Weight
Manufacturer
Melting Point (°C)
Product Line
Purity
Format
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing