![Staurosporine VETRANAL®, analytical standard](/deepweb/assets/sigmaaldrich/product/structures/320/336/3f7d7265-880c-4f97-b4a2-9fbdfd75a5d6/640/3f7d7265-880c-4f97-b4a2-9fbdfd75a5d6.png)
Staurosporine
![Staurosporine VETRANAL®, analytical standard](/deepweb/assets/sigmaaldrich/product/structures/320/336/3f7d7265-880c-4f97-b4a2-9fbdfd75a5d6/640/3f7d7265-880c-4f97-b4a2-9fbdfd75a5d6.png)
Synonym(s):
Antibiotic AM-2282
Empirical Formula (Hill Notation):
C28H26N4O3
CAS Number:
Molecular Weight:
466.53
Products
Product Category
Brand
Biological Source
Formula Weight
Physical Form
Product Line
Purity
Activity Spectrum
Format
Quality Segment
Application
Mode of Action
Reference Material Type
Special Grade
Technique