![Staphylococcal enterotoxin B from Staphylococcus aureus](/deepweb/assets/sigmaaldrich/product/images/350/166/e8fab925-564e-464f-993a-dee8be4260ef/640/e8fab925-564e-464f-993a-dee8be4260ef.jpg)
Staphylococcal enterotoxin B from Staphylococcus aureus
![Staphylococcal enterotoxin B from Staphylococcus aureus](/deepweb/assets/sigmaaldrich/product/images/350/166/e8fab925-564e-464f-993a-dee8be4260ef/640/e8fab925-564e-464f-993a-dee8be4260ef.jpg)
Synonym(s):
Enterotoxin B, Staphylococcal, SEB
CAS Number:
Products
Product Category
Brand
Chemical Composition
Quality Segment
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing