![N-(2,6-Dichlorophenyl)-2-indolinone British Pharmacopoeia (BP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/280/972/76604bd2-921f-434e-90e7-8a3c4a429d7c/640/76604bd2-921f-434e-90e7-8a3c4a429d7c.png)
Diclofenac related compound A
![N-(2,6-Dichlorophenyl)-2-indolinone British Pharmacopoeia (BP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/280/972/76604bd2-921f-434e-90e7-8a3c4a429d7c/640/76604bd2-921f-434e-90e7-8a3c4a429d7c.png)
Synonym(s):
N-(2,6-Dichlorophenyl)-2-indolinone, 1-(2,6-Dichlorophenyl)-1,3-dihydro-2H-indol-2-one, NSC 621845
Empirical Formula (Hill Notation):
C14H9Cl2NO
Products
Brand
Formula Weight
Manufacturer
Physical Form
Purity
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique
Showing 1-5 of 5
Compare
Product Number
Description
Pricing