Skip to Content
AR
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Stainless Steel - AISI 310 alloy, FeCr25Ni20
Products
Manufacturer
Physical Form
Diameter (mm)
Length (mm)
Filter Products
Showing 1-20 of 22
Page 1 of 2
Page 1 of 2
Compare
Product Number
Description
Pricing
GF33451365
Stainless Steel - AISI 310 alloy, FeCr25Ni20,
100mm rod, 12.7mm diameter, annealed
Expand
View Pricing
GF54180794
Stainless Steel - AISI 310 alloy, FeCr25Ni20,
200mm rod, 19.0mm diameter, annealed
Expand
View Pricing
GF74913374
Stainless Steel - AISI 310 alloy, FeCr25Ni20,
1000mm rod, 19.0mm diameter, annealed
Expand
View Pricing
GF03105100
Stainless Steel - AISI 310 alloy, FeCr25Ni20,
100mm rod, 6.35mm diameter, annealed
Expand
View Pricing
GF88137940
Stainless Steel - AISI 310 alloy, FeCr25Ni20,
200mm rod, 6.35mm diameter, annealed
Expand
View Pricing
GF92259467
Stainless Steel - AISI 310 alloy, FeCr25Ni20,
1000mm rod, 12.7mm diameter, annealed
Expand
View Pricing
GF72599305
Stainless Steel - AISI 310 alloy, FeCr25Ni20,
500mm rod, 19.0mm diameter, annealed
Expand
View Pricing
GF61643527
Stainless Steel - AISI 310 alloy, FeCr25Ni20,
foil, 300 x 300mm, 2.0mm thickness, annealed
Expand
View Pricing
GF71733742
Stainless Steel - AISI 310 alloy, FeCr25Ni20,
foil, 50 x 50mm, 2.0mm thickness, annealed
Expand
View Pricing
GF81420308
Stainless Steel - AISI 310 alloy, FeCr25Ni20,
200mm rod, 12.7mm diameter, annealed
Expand
View Pricing
GF39693145
Stainless Steel - AISI 310 alloy, FeCr25Ni20,
foil, 150 x 150mm, 2.0mm thickness, annealed
Expand
View Pricing
GF35171018
Stainless Steel - AISI 310 alloy, FeCr25Ni20,
foil, 150 x 150mm, 0.914mm thickness, annealed
Expand
View Pricing
GF87195853
Stainless Steel - AISI 310 alloy, FeCr25Ni20,
foil, 50 x 50mm, 0.914mm thickness, annealed
Expand
View Pricing
GF32433443
Stainless Steel - AISI 310 alloy, FeCr25Ni20,
100mm rod, 19.0mm diameter, annealed
Expand
View Pricing
GF17243748
Stainless Steel - AISI 310 alloy, FeCr25Ni20,
500mm rod, 6.35mm diameter, annealed
Expand
View Pricing
GF66717546
Stainless Steel - AISI 310 alloy, FeCr25Ni20,
foil, 100 x 100mm, 2.0mm thickness, annealed
Expand
View Pricing
GF96376977
Stainless Steel - AISI 310 alloy, FeCr25Ni20,
foil, 300 x 300mm, 0.914mm thickness, annealed
Expand
View Pricing
GF99481360
Stainless Steel - AISI 310 alloy, FeCr25Ni20,
foil, 100 x 100mm, 0.914mm thickness, annealed
Expand
View Pricing
GF77538077
Stainless Steel - AISI 310 alloy, FeCr25Ni20,
500mm rod, 12.7mm diameter, annealed
Expand
View Pricing
GF55467614
Stainless Steel - AISI 310 alloy, FeCr25Ni20,
1000mm rod, 6.35mm diameter, annealed
Expand
View Pricing
Page 1 of 2
Page 1 of 2