![Spiromesifen Metabolite M01 PESTANAL®, analytical standard](/deepweb/assets/sigmaaldrich/product/structures/218/554/c0e02408-6c0b-4fd5-b4dc-510664a76eb0/640/c0e02408-6c0b-4fd5-b4dc-510664a76eb0.png)
Spiromesifen Metabolite M01
![Spiromesifen Metabolite M01 PESTANAL®, analytical standard](/deepweb/assets/sigmaaldrich/product/structures/218/554/c0e02408-6c0b-4fd5-b4dc-510664a76eb0/640/c0e02408-6c0b-4fd5-b4dc-510664a76eb0.png)
Synonym(s):
4-Hydroxy-3-mesityl-1-oxaspiro(4.4)non-3-en-2-one
Empirical Formula (Hill Notation):
C17H20O3
CAS Number:
Molecular Weight:
272.34
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2