Skip to Content
Merck
Sodium thiosulfate United States Pharmacopeia (USP) Reference Standard

Sodium thiosulfate

Sodium thiosulfate United States Pharmacopeia (USP) Reference Standard
Synonym(s):
Sodium thiosulfate pentahydrate
Linear Formula:
Na2S2O3 · 5H2O
CAS Number:
Molecular Weight:
248.18

Products

Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade