Skip to Content
AR
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Sodium sulfate anhydrous
Synonym(s):
Sodium sulfate
Linear Formula:
Na
2
SO
4
CAS Number:
7757-82-6
Molecular Weight:
142.04
Products
Product Category
salts (2)
Brand
SAFC (2)
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
API Family
Special Grade
pharmaceutical primary standard (1)
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
1.28211
Sodium sulfate anhydrous,
EMPROVE
®
ESSENTIAL, Ph. Eur., BP, ChP, USP, E 514
Expand
View Pricing
1614807
Sodium sulfate anhydrous,
United States Pharmacopeia (USP) Reference Standard
Expand
View Pricing
1.28225
Sodium sulfate anhydrous,
EMPROVE
®
EXPERT, Ph. Eur., BP, ChP, USP
Expand
View Pricing