Sodium Stearyl Fumarate
Synonym(s):
Sodium stearyl fumarate, (E)-2-Butenedioic acid monooctadecyl ester sodium salt, Sodium octadecyl fumarate
Empirical Formula (Hill Notation):
C22H39NaO4
CAS Number:
Molecular Weight:
390.53
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2