Skip to Content
Merck
Sodium p-toluenesulfinate hydrate 97%

Sodium p-toluenesulfinate hydrate

Sodium p-toluenesulfinate hydrate 97%
Synonym(s):
p-Toluenesulfinic acid sodium salt hydrate
Linear Formula:
4-CH3C6H4SO2Na · xH2O
CAS Number:
Molecular Weight:
178.18 (anhydrous basis)
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing