Skip to Content
Merck
Sodium phosphate monobasic dihydrate purum p.a., crystallized, ≥99.0% (T)

Sodium phosphate monobasic dihydrate

Sodium phosphate monobasic dihydrate purum p.a., crystallized, ≥99.0% (T)
Synonym(s):
Sodium dihydrogen phosphate dihydrate
Linear Formula:
NaH2PO4 · 2H2O
CAS Number:
Molecular Weight:
156.01
EC Number:

Products

Product Category
Brand
Feature
Formula Weight
pH Value
Physical Form
Product Line
Purity
Quality Segment
Application
Special Grade