Skip to Content
Merck
Sodium perborate tetrahydrate purum p.a., 96% (RT)

Sodium perborate tetrahydrate

Sodium perborate tetrahydrate purum p.a., 96% (RT)
Synonym(s):
Sodium perborate tetrahydrate
Linear Formula:
NaBO3 · 4H2O
CAS Number:
Molecular Weight:
153.86

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Special Grade
Showing 1-3 of 3