![Sodium glyoxylate monohydrate ≥93%](/deepweb/assets/sigmaaldrich/product/structures/340/535/7ac44002-25da-482a-854b-8d22098cedc4/640/7ac44002-25da-482a-854b-8d22098cedc4.png)
Sodium glyoxylate monohydrate
![Sodium glyoxylate monohydrate ≥93%](/deepweb/assets/sigmaaldrich/product/structures/340/535/7ac44002-25da-482a-854b-8d22098cedc4/640/7ac44002-25da-482a-854b-8d22098cedc4.png)
Synonym(s):
Glyoxylic acid sodium salt monohydrate, Oxoethanoic acid sodium salt
Linear Formula:
HC(O)COONa · H2O
CAS Number:
Molecular Weight:
114.03
Products
Product Category
Brand
Formula Weight
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing